Learn More
Apexbio Technology LLC MDV3100 (Enzalutamide), 915087-33-1, MFCD14155804, 50 mg
SDP
Catalog No. 501013980
Encompass_Preferred
MF: C21H16F4N4O2S, MW: 464.4, Synonyms: Enzalutamide, MDV3100, MDV-3100, MDV 3100. Solubility: 23.22mg/mL in DMSO. MDV3100, known as Enzalutamide, is a second-generation androgen receptor (AR) signaling inhibitor. It has been demonstrated impressive affinity to the AR compared to the first-generation AR inhibitors. It is able to inhibit binding of androgens to the AR, AR nuclear translocation, and the association of the AR with DNA.
Catalog No. 50-101-3980 Supplier Apexbio Technology LLC Supplier No. A300350
MF: C21H16F4N4O2S, MW: 464.4, Synonyms: Enzalutamide, MDV3100, MDV-3100, MDV 3100. Solubility: 23.22mg/mL in DMSO. MDV3100, known as Enzalutamide, is a second-generation androgen receptor (AR) signaling inhibitor. It has been demonstrated impressive affinity to the AR compared to the first-generation AR inhibitors. It is able to inhibit binding of androgens to the AR, AR nuclear translocation, and the association of the AR with DNA.