Learn More
Adam Equipment MDW 300L Health Scale, Digital with height rod, Capacity: 660lb / 300kg with readability of 0.1lb / 0.05kg
Catalog No. NC2611804
Encompass_Preferred
Designed for easy operation, Adam Equipment's MDW digital scales meet the needs of medical professionals and health practitioners. MDW digital scales feature an adjustable height rod to provide simultaneous height and weight readings, and some models also provide body mass index (BMI) measurements. All MDW digital scales feature a built-in battery and easy-to-use, color-coded keypad. To ensure stability and patient security, MDW digital scales feature rugged construction with a sturdy metal base and a non-slip weighing platform.BMI function can calculate body mass indexThe MDW 300L offeres RS-232 interface for fast communication with printers and computersFeatures a hold function to lock in the weight on the displayInculded AC adapter and rechargeable batteryHeight measurements from 60cm to 212cmIntegrated wheels permit easy movement Maximum weighing capcity of 660lb / 300kgDisplay reads to 0.1lb / 0.05kg
Catalog No. NC2611804 Supplier Adam Equipment Supplier No. MDW 300L
Designed for easy operation, Adam Equipment's MDW digital scales meet the needs of medical professionals and health practitioners. MDW digital scales feature an adjustable height rod to provide simultaneous height and weight readings, and some models also provide body mass index (BMI) measurements. All MDW digital scales feature a built-in battery and easy-to-use, color-coded keypad. To ensure stability and patient security, MDW digital scales feature rugged construction with a sturdy metal base and a non-slip weighing platform.BMI function can calculate body mass indexThe MDW 300L offeres RS-232 interface for fast communication with printers and computersFeatures a hold function to lock in the weight on the displayInculded AC adapter and rechargeable batteryHeight measurements from 60cm to 212cmIntegrated wheels permit easy movement Maximum weighing capcity of 660lb / 300kgDisplay reads to 0.1lb / 0.05kg