Learn More
Medchemexpress LLC ME-344 | 1374524-68-1 | 99.8% | 348.39 | 10 MG
SDP
Catalog No. 5000437043
Encompass_Preferred
ME-344 is an Isoflavone that increases mitochondrial ROS generation, inhibits tubulin polymerization, inhibits HO-1 and impacts its mitochondrial translocation, and induces Apoptosis through Caspase 3 activation. It synergizes with Vinblastine in leukemia cells and displays anti-tumor activity against leukemia and lung tumors. This compound can be used in the research of lung cancer, acute myeloid leukemia, and HER2-negative breast cancer.
  • Increases mitochondrial ROS generation.
  • Inhibits tubulin polymerization.
  • Inhibits HO-1 and impacts its mitochondrial translocation.
  • Induces apoptosis through Caspase 3 activation.
  • Synergizes with Vinblastine in leukemia cells.
  • Displays anti-tumor activity against leukemia and lung tumors.
  • Useful in research for lung cancer, acute myeloid leukemia, and HER2-negative breast cancer.
Catalog No. 50-004-37043 Supplier Medchemexpress LLC Supplier No. HY11274910MG
ME-344 is an Isoflavone that increases mitochondrial ROS generation, inhibits tubulin polymerization, inhibits HO-1 and impacts its mitochondrial translocation, and induces Apoptosis through Caspase 3 activation. It synergizes with Vinblastine in leukemia cells and displays anti-tumor activity against leukemia and lung tumors. This compound can be used in the research of lung cancer, acute myeloid leukemia, and HER2-negative breast cancer.
  • Increases mitochondrial ROS generation.
  • Inhibits tubulin polymerization.
  • Inhibits HO-1 and impacts its mitochondrial translocation.
  • Induces apoptosis through Caspase 3 activation.
  • Synergizes with Vinblastine in leukemia cells.
  • Displays anti-tumor activity against leukemia and lung tumors.
  • Useful in research for lung cancer, acute myeloid leukemia, and HER2-negative breast cancer.