Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Me-tet-peg4-cooh | 2141976-30-7 | 98.7% | 477.51 g·mol⁻¹ | C22H31N5O7 | 10 MG
SDP

Catalog No. 5000204394
Encompass_Preferred

Me-Tet-PEG4-COOH is an ADC linker containing four polyethylene glycol (PEG) units and a tetrazine group that enables inverse electron demand Diels-Alder (iEDDA) bioorthogonal conjugation with TCO-containing partners. Intended for research use in bioconjugation and ADC development.

  • Purity: 98.68%
  • Molecular weight: 477.51 g·mol⁻¹
  • Chemical formula: C22H31N5O7
  • Cas number: 2141976-30-7
  • Appearance: pale purple to purple solid
  • Solubility: DMSO 125 mg/mL (requires ultrasonic; use newly opened DMSO)
  • Storage: powder -20°C up to 3 years; in solvent -80°C for 6 months, -20°C for 1 month
  • Product type: ADC linker

Catalog No. 50-002-04394 Supplier Medchemexpress LLC Supplier No. HY15649210MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Me-Tet-PEG4-COOH is an ADC linker containing four polyethylene glycol (PEG) units and a tetrazine group that enables inverse electron demand Diels-Alder (iEDDA) bioorthogonal conjugation with TCO-containing partners. Intended for research use in bioconjugation and ADC development.

  • Purity: 98.68%
  • Molecular weight: 477.51 g·mol⁻¹
  • Chemical formula: C22H31N5O7
  • Cas number: 2141976-30-7
  • Appearance: pale purple to purple solid
  • Solubility: DMSO 125 mg/mL (requires ultrasonic; use newly opened DMSO)
  • Storage: powder -20°C up to 3 years; in solvent -80°C for 6 months, -20°C for 1 month
  • Product type: ADC linker

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.