Learn More
Medchemexpress LLC Me-triacetyl-β-D-glucopyranuronate-Ph-CH2OH-Fmoc | 894096-02-7 | 98.8% | 748.73 | C38H40N2O14 | 10 MG
SDP
Catalog No. 5000203710
Encompass_Preferred
Me-triacetyl-β-D-glucopyranuronate-Ph-CH2OH-Fmoc is a cleavable linker reagent used to assemble cleavable linker-payload constructs during the synthesis of antibody-drug conjugates (ADCs).
  • Cleavable glycoside-based linker for controlled payload release.
  • Fmoc-protected benzyl alcohol handle for selective conjugation.
  • Reported high purity suitable for research use (98.8%).
  • Available in small-scale quantities for medicinal chemistry and ADC development.
Catalog No. 50-002-03710 Supplier Medchemexpress LLC Supplier No. HY13108710MG
Me-triacetyl-β-D-glucopyranuronate-Ph-CH2OH-Fmoc is a cleavable linker reagent used to assemble cleavable linker-payload constructs during the synthesis of antibody-drug conjugates (ADCs).
  • Cleavable glycoside-based linker for controlled payload release.
  • Fmoc-protected benzyl alcohol handle for selective conjugation.
  • Reported high purity suitable for research use (98.8%).
  • Available in small-scale quantities for medicinal chemistry and ADC development.