Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Me-triacetyl-β-D-glucopyranuronate-Ph-CH2OH-Fmoc | 894096-02-7 | MFCD34470435 | 98.8% | 748.73 g/mol | C38H40N2O14 | 5 MG
SDP

Catalog No. 5000202982
Encompass_Preferred

Me-triacetyl-β-D-glucopyranuronate-Ph-CH2OH-Fmoc is a cleavable ADC linker used in the synthesis of antibody-drug conjugates (ADCs). It is supplied as a white to light yellow solid for research applications and is typically prepared as a DMSO stock for conjugation workflows.

  • Cleavable ADC linker for antibody-drug conjugation.
  • White to light yellow solid supplied in small research quantities.
  • Purity 98.8% (typical; batch dependent).
  • Soluble in DMSO at 125 mg/mL; ultrasonic agitation recommended for dissolution.
  • Store powder at -20°C; store solutions at -80°C for extended stability.

Catalog No. 50-002-02982 Supplier Medchemexpress LLC Supplier No. HY1310875MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Me-triacetyl-β-D-glucopyranuronate-Ph-CH2OH-Fmoc is a cleavable ADC linker used in the synthesis of antibody-drug conjugates (ADCs). It is supplied as a white to light yellow solid for research applications and is typically prepared as a DMSO stock for conjugation workflows.

  • Cleavable ADC linker for antibody-drug conjugation.
  • White to light yellow solid supplied in small research quantities.
  • Purity 98.8% (typical; batch dependent).
  • Soluble in DMSO at 125 mg/mL; ultrasonic agitation recommended for dissolution.
  • Store powder at -20°C; store solutions at -80°C for extended stability.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.