Learn More
Medchemexpress LLC HY-100225 5mg Medchemexpress, ME0328 CAS:1445251-22-8 Purity:>98%
SDP
Catalog No. 501931763
Encompass_Preferred
Medchemexpress, HY-100225 5mg ME0328 CAS:1445251-22-8 ME0328 is a potent and selective ARTD3 / PARP3 inhibitor with an IC 50 of 0.89±0.28 μM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-1763 Supplier Medchemexpress LLC Supplier No. HY1002255MG
Medchemexpress, HY-100225 5mg ME0328 CAS:1445251-22-8 ME0328 is a potent and selective ARTD3 / PARP3 inhibitor with an IC 50 of 0.89±0.28 μM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.