Learn More
Medchemexpress LLC Me1111 | 1391758-52-3 | 100.0% | 202.25 g/mol | C12H14N2O | 100 MG
SDP
Catalog No. 5000242450
Encompass_Preferred
ME1111 is an antifungal small molecule (CAS 1391758-52-3) that inhibits succinate dehydrogenase in Trichophyton species. It is active against dermatophytes and is provided for research use in antifungal and onychomycosis studies, with validated solubility and storage guidelines for in vitro and in vivo applications.
  • Antifungal activity against dermatophytes.
  • Inhibits succinate dehydrogenase in Trichophyton species.
  • High reported purity (99.95%).
  • Soluble in DMSO at 100 mg/mL for in vitro use.
  • Powder storage: -20°C for long-term stability.
Catalog No. 50-002-42450 Supplier Medchemexpress LLC Supplier No. HY108012100MG
ME1111 is an antifungal small molecule (CAS 1391758-52-3) that inhibits succinate dehydrogenase in Trichophyton species. It is active against dermatophytes and is provided for research use in antifungal and onychomycosis studies, with validated solubility and storage guidelines for in vitro and in vivo applications.
  • Antifungal activity against dermatophytes.
  • Inhibits succinate dehydrogenase in Trichophyton species.
  • High reported purity (99.95%).
  • Soluble in DMSO at 100 mg/mL for in vitro use.
  • Powder storage: -20°C for long-term stability.