Learn More
AdipoGen Me7BIO (5 mg)
SDP
Supplier:  AdipoGen AGMRC0022M005
Encompass_Preferred
Chemical. CAS: 916440-98-7. Formula: C17H12BrN3O2. MW: 370.2. N-methylated inactive control compound for 7BIO (Prod. No. MR-C0020). Aryl hydrocarbon receptor (AhR) agonist; causes redistribution of AhR to the nucleus.
Catalog No. 50-205-4243