Learn More
Enzo Life Sciences Mead acid (1mg). CAS: 20590-32-3
SDP
Supplier:  Enzo Life Sciences BMLFA0040001
Encompass_Preferred
Accumulates in tissues of essential fatty acid deficient animals. Is not converted to prostaglandins and LTB4 but is converted to cysteinyl leukotrienes. Immunomodulatory and anti-inflammatory effects. Purity: ≥97% (HPLC). Solubility: Soluble in ethanol, DMSO, acetone, ethyl acetate or hexane. Long Term Storage: -20°C.
Catalog No. 50-201-0795