Learn More
ABclonal Technology MEAF6 Rabbit pAb
SDP
Catalog No. 5000107729
Encompass_Preferred
This gene encodes a nuclear protein involved in transcriptional activation. The encoded protein may form a component of several different histone acetyltransferase complexes. There is a pseudogene for this gene on chromosome 2. Alternative splicing results in multiple transcript variants.
Catalog No. 50-001-07729 Supplier ABclonal Technology Supplier No. A14629500UL
This gene encodes a nuclear protein involved in transcriptional activation. The encoded protein may form a component of several different histone acetyltransferase complexes. There is a pseudogene for this gene on chromosome 2. Alternative splicing results in multiple transcript variants.