Learn More
Cayman Chemical MEAI (hydrochloride), Purity: ≥95%, CAS: 81593-54-6, 10 mg
SDP
Catalog No. 502968313
Encompass_Preferred
A derivative of MMAI, a psychoactive aminoindane that has been identified in recreational drug mixtures; intended for forensic and research purposes
Catalog No. 50-296-8313 Supplier Cayman Chemical Supplier No. 1828610 MG
A derivative of MMAI, a psychoactive aminoindane that has been identified in recreational drug mixtures; intended for forensic and research purposes