Learn More
MSC Bell Laboratories Meal Bait, 25 lb, Bromadiolone, for Rats & Mice
Catalog No. 502837959
Encompass
This rodenticide meal bait is designed to control infestations of rats and mice. Formulated with food-grade ingredients, grains, and seeds, it uses bromadiolone as its active ingredient to ensure a lethal dose can be consumed in a single feeding, leading to elimination within four to five days.
  • Controls mice and rats, including warfarin-resistant Norway rats.
  • Features a palatable granular formulation with food-grade ingredients, seeds, and grains.
  • Rodents can consume a lethal dose in one feeding.
  • Eliminates rodents in four to five days after consumption.
  • Suitable for sensitive areas where rodents might transport bait.
  • Can be used indoors and outdoors within tamper-resistant bait stations.
  • Effective against various pests, including multiple rat and mouse species.
  • Has a shelf life of up to 3 years when stored properly.
Catalog No. 50-283-7959 Supplier MSC Supplier No. 74313842
This rodenticide meal bait is designed to control infestations of rats and mice. Formulated with food-grade ingredients, grains, and seeds, it uses bromadiolone as its active ingredient to ensure a lethal dose can be consumed in a single feeding, leading to elimination within four to five days.
  • Controls mice and rats, including warfarin-resistant Norway rats.
  • Features a palatable granular formulation with food-grade ingredients, seeds, and grains.
  • Rodents can consume a lethal dose in one feeding.
  • Eliminates rodents in four to five days after consumption.
  • Suitable for sensitive areas where rodents might transport bait.
  • Can be used indoors and outdoors within tamper-resistant bait stations.
  • Effective against various pests, including multiple rat and mouse species.
  • Has a shelf life of up to 3 years when stored properly.