Learn More
Bioworld Measles Virus Large Polymerase (58-149) (Recombinant), 100 μg
Supplier:  Bioworld 220605531
Encompass_Preferred
Recombinant Measles Virus Large structural protein RNA Polymerase (58-149) Source: E.Coli Background: Description: E.coli derived recombinant. The protein contains the large polymerase immunodominant regions,amino acids: (58-149) Purification method: Purified by proprietary chromatographic technique. Purity: Protein is >95% pure as determined by 10% PAGE (coomassie staining) and RP-HPLC. Protein concentration: 1 mg/ml containing 50mM Tris-HCl, pH 8.0, 60mM NaCl, 10mM glutathione, 0.1% sarcosil and 50% glycero1. Storage: Protein is shipped at ambient temperature. Upon arrival, Store at -200C. Stability: Five years frozen. One month in solution at room temperature. Application: Use as an antigen in ELISA and Western Blots. Immunoreactive with sera of Measles Virus infected individuals. Usage: BioWORLD\'s products are furnished for LABORATORY RESEARCH USE ONLY. The product may not be used as drugs, agricultural or pesticidal products, food additives or household chemicals.
Catalog No. 50-253-652