Learn More
ATCC Measles virus RNA; Strain: Edmonston
Catalog No. 502382958
Encompass
Genomic RNA from the preparation of cell lysate and supernatant from MRC-5 cells (ATCC CCL-171) infected with the measles virus strain Edmonston (ATCC VR-24). This product was prepared using methods known to inactivate viruses. It is suitable for use in RT-PCR or other RNA-based procedures. Source organisms and host cells are also available through the ATCC Catalog.
Catalog No. 50-238-2958 Supplier ATCC Supplier No. VR24D
Genomic RNA from the preparation of cell lysate and supernatant from MRC-5 cells (ATCC CCL-171) infected with the measles virus strain Edmonston (ATCC VR-24). This product was prepared using methods known to inactivate viruses. It is suitable for use in RT-PCR or other RNA-based procedures. Source organisms and host cells are also available through the ATCC Catalog.