Learn More
Bioss Measles virus- Rubeola
Supplier: Bioss BSM72310M
Measles virus(Rubeola) Primary Antibody Unconjugated Host Species Mouse Target Species Clonality Monoclonal Applications ELISA IF Concentration 100 g/ml Size 100 ug Storage Buffer PBS (pH 7 2) with 0 1% azide