Learn More
Bioss Measles virus - Rubeola
SDP
Catalog No. 5000065571
Encompass_Preferred
Measles virus(Rubeola) Primary Antibody Unconjugated Host Species Mouse Target Species Clonality Monoclonal Applications ELISA IF Concentration 100 g/ml Size 100 ug Storage Buffer PBS (pH 7 2) with 0 1% azide
Catalog No. 50-000-65571 Supplier Bioss Supplier No. BSM72307M
Measles virus(Rubeola) Primary Antibody Unconjugated Host Species Mouse Target Species Clonality Monoclonal Applications ELISA IF Concentration 100 g/ml Size 100 ug Storage Buffer PBS (pH 7 2) with 0 1% azide