Learn More
ATCC Measles virus strain Edmonston (ATCC VR-24™)
Catalog No. 502383270
Encompass
Measles virus strain Edmonston (ATCC VR-24™) is a whole-genome sequenced prototype strain isolated in 1954 from a human patient in Massachusetts. Propagated in MRC-5 cells, it is used in respiratory and infectious disease research.
Recommended companion products:
MRC-5 cells (Fisher Cat. No. 50-238-3231, ATCC CCL-171™)
EMEM (Fisher Cat. No. 50-238-2632, ATCC 30-2003™)
Fetal bovine serum (Fisher Cat. No. 50-238-3771, ATCC 30-2020™).
Catalog No. 50-238-3270 Supplier ATCC Supplier No. VR24
 Restricted Item
Add to Cart
Add to Cart
Measles virus strain Edmonston (ATCC VR-24™) is a whole-genome sequenced prototype strain isolated in 1954 from a human patient in Massachusetts. Propagated in MRC-5 cells, it is used in respiratory and infectious disease research.
Recommended companion products:
MRC-5 cells (Fisher Cat. No. 50-238-3231, ATCC CCL-171™)
EMEM (Fisher Cat. No. 50-238-2632, ATCC 30-2003™)
Fetal bovine serum (Fisher Cat. No. 50-238-3771, ATCC 30-2020™).