Learn More
Electron Microscopy Sciences Measure-Matic™ Dispenser
SDP
Catalog No. 501926886
Encompass_Preferred
Measuring dispenser makes repeated measuring and pouring easy. Adjust the dispensing cup for volumes from 5 ml to 25 ml, then squeeze the 500 ml reservoir bottle. Reservoir bottle is made of squeezable-density polyethylene. Dust cover included. Cup and closure are made of rugged polypropylene. Reservoir bottle is made of squeezable-density polyethylene.
Catalog No. 50-192-6886 Supplier Electron Microscopy Sciences Supplier No. 6429908
Measuring dispenser makes repeated measuring and pouring easy. Adjust the dispensing cup for volumes from 5 ml to 25 ml, then squeeze the 500 ml reservoir bottle. Reservoir bottle is made of squeezable-density polyethylene. Dust cover included. Cup and closure are made of rugged polypropylene. Reservoir bottle is made of squeezable-density polyethylene.