Learn More
Grainger Measure Miser 11.88 in L Solid PK12
Catalog No. 502751903
Encompass_Preferred
The Carlisle Measure Miser 11-88 in. L Solid Phenolic Partition is an 11-inch wide by 88-inch long panel made from durable, moisture-resistant, and fire-resistant phenolic material. It has a Class A fire rating, low smoke development, and low flame spread. The panel features tongue and groove edges for easy assembly and can be cut to size. It resists mold, mildew, and bacteria growth, making it ideal for healthcare, laboratory, industrial, or office environments where fire resistance, durability, and cleanability are essential.
Catalog No. 50-275-1903 Supplier Grainger Supplier No. 14D256
The Carlisle Measure Miser 11-88 in. L Solid Phenolic Partition is an 11-inch wide by 88-inch long panel made from durable, moisture-resistant, and fire-resistant phenolic material. It has a Class A fire rating, low smoke development, and low flame spread. The panel features tongue and groove edges for easy assembly and can be cut to size. It resists mold, mildew, and bacteria growth, making it ideal for healthcare, laboratory, industrial, or office environments where fire resistance, durability, and cleanability are essential.