Learn More
Chemglass Life Sciences Cylinder, Class B, 250mL, Graduated, Single Scale
SDP
Catalog No. NC1907760
Encompass_Preferred
Polyethylene bumper guards are supplied with sizes 25 through 2000mL. Scale is durable white ceramic enamel. Designed from ASTM Specification E1272, Style 1, Class B requirements.
Catalog No. NC1907760 Supplier Chemglass Life Sciences Supplier No. CG8242250
Polyethylene bumper guards are supplied with sizes 25 through 2000mL. Scale is durable white ceramic enamel. Designed from ASTM Specification E1272, Style 1, Class B requirements.