Learn More
Foxx Life Sciences Borosil® Class B TC Graduated Measuring Cylinder with Pour Spout, Hexagonal Base, and Permanent Amber Graduations, 5 mL, 5/CS
Catalog No. 501720379
Encompass_Preferred
Each Borosil® Class B TC Cylinder is calibrated by automatic laser calibration. Certificates can be downloaded by visiting Borosil.com, or by downloading the Borosil app. Borosil® Cylinders are manufactured with hexagonal, spout-aligned bases for improved stability. Permanent amber graduations will not fade, even after years of use. Available from 5mL to 2L. Cylinders comply with ASTM E-1272. In collaboration with Borosil Glass Works, Foxx Life Sciences is proud to bring the outstanding quality of Borosil® Type 3.3 Borosilicate glassware to North America for the first time. Manufactured from the same USP Type 1 borosilicate glass that North American scientists have come to trust, Borosil® Laboratory Glassware offers unparalleled mechanical strength, clarity, and reliability at competitive prices. And thanks to a unique annealing process, Borosil® Glassware is stress-free and incredibly resistant to thermal shock.
Catalog No. 50-172-0379 Supplier Foxx Life Sciences Supplier No. 3027005
Each Borosil® Class B TC Cylinder is calibrated by automatic laser calibration. Certificates can be downloaded by visiting Borosil.com, or by downloading the Borosil app. Borosil® Cylinders are manufactured with hexagonal, spout-aligned bases for improved stability. Permanent amber graduations will not fade, even after years of use. Available from 5mL to 2L. Cylinders comply with ASTM E-1272. In collaboration with Borosil Glass Works, Foxx Life Sciences is proud to bring the outstanding quality of Borosil® Type 3.3 Borosilicate glassware to North America for the first time. Manufactured from the same USP Type 1 borosilicate glass that North American scientists have come to trust, Borosil® Laboratory Glassware offers unparalleled mechanical strength, clarity, and reliability at competitive prices. And thanks to a unique annealing process, Borosil® Glassware is stress-free and incredibly resistant to thermal shock.