Learn More
Foxx Life Sciences Borosil Class B TC Graduated Measuring Cylinder with Pour Spout, Hexagonal Base, and Permanent Amber Graduations, Individual Certificate, 5mL, 5/CS
Catalog No. 501720286
Encompass_Preferred
Borosil, Graduated Measuring Cylinder, Pour Out, Single Metric, Class A Accuracy as per ASTM E 1272, with Individual Certificate, with Hexagonal Base, 5mL, C/S 5Borosil Measuring Cylinders comply with ASTM E 1272. Calibrated as per Class A tolerances, each cylinder is engraved with a unique serial number and comes with a Certificate of Calibration traceable to national standards. The graduations are marked with Amber enamel and the Cylinder is manufactured with chemically resistant 3.3 borosilicate glass. Its sturdy hexagonal base prevents roll-over issues and ensures for better stability. Comes with Individual Certificate.
Catalog No. 50-172-0286 Supplier Foxx Life Sciences Supplier No. 3026005D
Borosil, Graduated Measuring Cylinder, Pour Out, Single Metric, Class A Accuracy as per ASTM E 1272, with Individual Certificate, with Hexagonal Base, 5mL, C/S 5Borosil Measuring Cylinders comply with ASTM E 1272. Calibrated as per Class A tolerances, each cylinder is engraved with a unique serial number and comes with a Certificate of Calibration traceable to national standards. The graduations are marked with Amber enamel and the Cylinder is manufactured with chemically resistant 3.3 borosilicate glass. Its sturdy hexagonal base prevents roll-over issues and ensures for better stability. Comes with Individual Certificate.