Learn More
Grainger Measuring Funnel 1 qt. 1-3/8 Dia Spout
Catalog No. 502751787
Encompass_Preferred
The Grainger 14N898 is a Westward brand 1/4-inch drive socket adapter that converts a 1/4-inch square drive to a 3/8-inch square drive. It is made from chrome vanadium steel for durability and has a black oxide finish to resist corrosion. This socket adapter allows you to use 1/4-inch drive tools with a 3/8-inch drive ratchet, extension bar, or other accessory. It measures 1-1/2 inches long and weighs 0.8 ounces. The 14N898 socket adapter is ideal for automotive applications or projects where you need to adapt between different drive sizes.
Catalog No. 50-275-1787 Supplier Grainger Supplier No. 14N898
The Grainger 14N898 is a Westward brand 1/4-inch drive socket adapter that converts a 1/4-inch square drive to a 3/8-inch square drive. It is made from chrome vanadium steel for durability and has a black oxide finish to resist corrosion. This socket adapter allows you to use 1/4-inch drive tools with a 3/8-inch drive ratchet, extension bar, or other accessory. It measures 1-1/2 inches long and weighs 0.8 ounces. The 14N898 socket adapter is ideal for automotive applications or projects where you need to adapt between different drive sizes.