Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Measuring Jars
SCI_ED

Catalog No. S95916
Change view
Click to view available options
Capacity (Metric):
3000 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
S95916 3000 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S95916 Supplier Learning Resources, Inc Supplier No. LER2462
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Perfect for both customary and metric measurements.

  • Set of five sturdy plastic jars includes screw-on covers
  • Customary capacities are gallon, half-gallon, quart, pint and gill
  • Metric measurements range from 100 to 3000mL
  • Grades 1-8

Specifications

Capacity (Metric) 3000 mL
Includes Screw On Lid
Material Plastic
Closure Type Screw Cap
Shape Round
Product Type Measuring Jug
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.