Learn More
Electron Microscopy Sciences Measuring Scale #1 for 10X Measuring Magnifier, 35 mm
SDP
Catalog No. 5027071
Encompass_Preferred
These scales are for use with the Measuring Magnifiers. Measuring scale for 10x magnifiers. Degrees: 0-90° (1° intervals). Circles: 0.1 to 3.0mm dia. Scale: ± 0 to 10mm (0.1mm divisions). Inch Scale: 0 to 0.4" (0.005" divisions). Size 35mm.
Use: 10X
Size: 35 mm
Catalog No. 50-270-71 Supplier Electron Microscopy Sciences Supplier No. 68162
These scales are for use with the Measuring Magnifiers. Measuring scale for 10x magnifiers. Degrees: 0-90° (1° intervals). Circles: 0.1 to 3.0mm dia. Scale: ± 0 to 10mm (0.1mm divisions). Inch Scale: 0 to 0.4" (0.005" divisions). Size 35mm.
Use: 10X
Size: 35 mm