Learn More
Electron Microscopy Sciences Measuring Scale #1 for 7X or 15X Measuring Magnifier, 26 mm
SDP
Catalog No. 5027083
Encompass_Preferred
These scales are for use with the Measuring Magnifiers. Measuring scale for 7x and 15x magnifiers. Degrees: 0-90° (1° intervals). Circles: 0.1 to 3.0mm dia. Scale: ± 0 to 10mm (0.1mm divisions). Inch Scale: 0 to 0.4" (0.005" divisions). Squares: 0.025". Size 26mm.
Use: 7X and 15X
Size: 26 mm
Catalog No. 50-270-83 Supplier Electron Microscopy Sciences Supplier No. 68161
These scales are for use with the Measuring Magnifiers. Measuring scale for 7x and 15x magnifiers. Degrees: 0-90° (1° intervals). Circles: 0.1 to 3.0mm dia. Scale: ± 0 to 10mm (0.1mm divisions). Inch Scale: 0 to 0.4" (0.005" divisions). Squares: 0.025". Size 26mm.
Use: 7X and 15X
Size: 26 mm