Learn More
Electron Microscopy Sciences Measuring Scale #10 for 10X Measuring Magnifier, 35 mm
SDP
Catalog No. 5027070
Encompass_Preferred
These scales are for use with the Measuring Magnifiers. Measuring scale for 10x magnifiers. Screw Threads: 0.25, 0.3, 0.35, 0.4, 0.6, 0.75, 0.9, 1mm.
Use: 10X
Size: 35 mm
Catalog No. 50-270-70 Supplier Electron Microscopy Sciences Supplier No. 68180
These scales are for use with the Measuring Magnifiers. Measuring scale for 10x magnifiers. Screw Threads: 0.25, 0.3, 0.35, 0.4, 0.6, 0.75, 0.9, 1mm.
Use: 10X
Size: 35 mm