Learn More
Electron Microscopy Sciences Measuring Scale #10 for 7X Measuring Magnifier, 26 mm
SDP
Catalog No. 5027065
Encompass_Preferred
These scales are for use with the Measuring Magnifiers. Measuring scale for 7x magnifiers. Screw Threads: 0.25, 0.3, 0.35, 0.4, 0.6, 0.75, 0.9, 1mm.
Use: 7X
Size: 26 mm
Catalog No. 50-270-65 Supplier Electron Microscopy Sciences Supplier No. 68179
These scales are for use with the Measuring Magnifiers. Measuring scale for 7x magnifiers. Screw Threads: 0.25, 0.3, 0.35, 0.4, 0.6, 0.75, 0.9, 1mm.
Use: 7X
Size: 26 mm