Learn More
Electron Microscopy Sciences Measuring Scale #11 for 10X Measuring Magnifier, 35 mm
SDP
Catalog No. 5027084
Encompass_Preferred
These scales are for use with the Measuring Magnifiers. Measuring scale for 10x magnifiers. Screw Threads: 50, 18, 20, 8, 16, 32, 24, 28, 14 per inch.
Use: 10X
Size: 35 mm
Catalog No. 50-270-84 Supplier Electron Microscopy Sciences Supplier No. 68182
These scales are for use with the Measuring Magnifiers. Measuring scale for 10x magnifiers. Screw Threads: 50, 18, 20, 8, 16, 32, 24, 28, 14 per inch.
Use: 10X
Size: 35 mm