Learn More
Electron Microscopy Sciences Measuring Scale #12 for 10X Measuring Magnifier, 35 mm
SDP
Catalog No. 5027086
Encompass_Preferred
These scales are for use with the Measuring Magnifiers. Measuring scale for 10x magnifiers. Degrees: 0-90° (1° intervals). Radius: 1/16 to 3/8" (1/32" divisions). Scales: 0 to .5" (0.005" divisions); 0 to 10mm (0.1mm divisions). Line Thickness: 0.01", 0.02", 0.03".
Use: 10X
Size: 35 mm
Catalog No. 50-270-86 Supplier Electron Microscopy Sciences Supplier No. 68184
These scales are for use with the Measuring Magnifiers. Measuring scale for 10x magnifiers. Degrees: 0-90° (1° intervals). Radius: 1/16 to 3/8" (1/32" divisions). Scales: 0 to .5" (0.005" divisions); 0 to 10mm (0.1mm divisions). Line Thickness: 0.01", 0.02", 0.03".
Use: 10X
Size: 35 mm