Learn More
Electron Microscopy Sciences Measuring Scale #12 for 7X Measuring Magnifier, 26 mm
SDP
Catalog No. 5027085
Encompass_Preferred
These scales are for use with the Measuring Magnifiers. Measuring scale for 7x magnifiers. Degrees: 0-90° (1° intervals). Radius: 1/16 to 3/8" (1/32" divisions). Scales: 0 to .5" (0.005" divisions); 0 to 10mm (0.1mm divisions). Line Thickness: 0.01", 0.02", 0.03".
Use: 7X
Size: 26 mm
Catalog No. 50-270-85 Supplier Electron Microscopy Sciences Supplier No. 68183
These scales are for use with the Measuring Magnifiers. Measuring scale for 7x magnifiers. Degrees: 0-90° (1° intervals). Radius: 1/16 to 3/8" (1/32" divisions). Scales: 0 to .5" (0.005" divisions); 0 to 10mm (0.1mm divisions). Line Thickness: 0.01", 0.02", 0.03".
Use: 7X
Size: 26 mm