Learn More
Electron Microscopy Sciences Measuring Scale #2 for 10X Measuring Magnifier, 35 mm
SDP
Catalog No. 5027076
Encompass_Preferred
These scales are for use with the Measuring Magnifiers. Measuring scale for 10x magnifiers. 35mm Degrees: 0-90° (0 to 10° marked in 1°) Circles: 0.1 to 1.0mm dia. And .005 to 1/16" dia. Scales: 0 to .5" (0.005" divisions); 0 to 1/8" (1/64" divisions); ± 0 to 10mm (0.1mm divisions). Size 35mm.
Use: 10X
Size: 35 mm
Catalog No. 50-270-76 Supplier Electron Microscopy Sciences Supplier No. 68164
These scales are for use with the Measuring Magnifiers. Measuring scale for 10x magnifiers. 35mm Degrees: 0-90° (0 to 10° marked in 1°) Circles: 0.1 to 1.0mm dia. And .005 to 1/16" dia. Scales: 0 to .5" (0.005" divisions); 0 to 1/8" (1/64" divisions); ± 0 to 10mm (0.1mm divisions). Size 35mm.
Use: 10X
Size: 35 mm