Learn More
Electron Microscopy Sciences Measuring Scale #3 for 7X Measuring Magnifier, 26 mm
SDP
Catalog No. 5027077
Encompass_Preferred
These scales are for use with the Measuring Magnifiers. Measuring scale for 7x magnifiers. Lines: 0.02 to 0.16mm thick. Size 26mm.
Use: 7X
Size: 26 mm
Catalog No. 50-270-77 Supplier Electron Microscopy Sciences Supplier No. 68165
These scales are for use with the Measuring Magnifiers. Measuring scale for 7x magnifiers. Lines: 0.02 to 0.16mm thick. Size 26mm.
Use: 7X
Size: 26 mm