Learn More
Electron Microscopy Sciences Measuring Scale #4 for 7X Measuring Magnifier, 26 mm
SDP
Catalog No. 5027069
Encompass_Preferred
These scales are for use with the Measuring Magnifiers. Measuring scale for 7x magnifiers. Degrees: 0-90° (one 10° area marked in 1°). Scale: ± 0 to 10mm (0.1mm div). Squares: 5 x 10mm (0.1mm div). Circles: 0.1 to 1.0mm dia. (0.1mm divisions). Radius: 0-90° (0.1 and 0.5mm div) 26mm.
Use: 7X
Size: 26 mm
Catalog No. 50-270-69 Supplier Electron Microscopy Sciences Supplier No. 68167
These scales are for use with the Measuring Magnifiers. Measuring scale for 7x magnifiers. Degrees: 0-90° (one 10° area marked in 1°). Scale: ± 0 to 10mm (0.1mm div). Squares: 5 x 10mm (0.1mm div). Circles: 0.1 to 1.0mm dia. (0.1mm divisions). Radius: 0-90° (0.1 and 0.5mm div) 26mm.
Use: 7X
Size: 26 mm