Learn More
Electron Microscopy Sciences Measuring Scale #5 for 10X Measuring Magnifier, 35 mm
SDP
Catalog No. 5027075
Encompass_Preferred
These scales are for use with the Measuring Magnifiers. Measuring scale 10x magnifier. Degrees: 0-90° (5° interval). Scales: 0 to 0.5" (0.005" div); 0 to 10mm (0.2mm). Circles: 1/64 to 1/16"; .002 to .050mm. Radius: 0-90° (1/16 to 3/8" at 1/32"). Line lengths: 1/64-1/4" (1/32").
Use: 10X
Size: 35 mm
Catalog No. 50-270-75 Supplier Electron Microscopy Sciences Supplier No. 68170
These scales are for use with the Measuring Magnifiers. Measuring scale 10x magnifier. Degrees: 0-90° (5° interval). Scales: 0 to 0.5" (0.005" div); 0 to 10mm (0.2mm). Circles: 1/64 to 1/16"; .002 to .050mm. Radius: 0-90° (1/16 to 3/8" at 1/32"). Line lengths: 1/64-1/4" (1/32").
Use: 10X
Size: 35 mm