Learn More
Electron Microscopy Sciences Measuring Scale #5 for 7X Measuring Magnifier, 26 mm
SDP
Catalog No. 5027064
Encompass_Preferred
These scales are for use with the Measuring Magnifiers. Measuring scale 7x magnifier. Degrees: 0-90° (5° interval). Scales: 0 to 0.5" (0.005" div); 0 to 10mm (0.2mm). Circles: 1/64 to 1/16"; .002 to .050mm. Radius: 0-90° (1/16 to 3/8" at 1/32").
Use: 7X
Size: 26 mm
Catalog No. 50-270-64 Supplier Electron Microscopy Sciences Supplier No. 68169
These scales are for use with the Measuring Magnifiers. Measuring scale 7x magnifier. Degrees: 0-90° (5° interval). Scales: 0 to 0.5" (0.005" div); 0 to 10mm (0.2mm). Circles: 1/64 to 1/16"; .002 to .050mm. Radius: 0-90° (1/16 to 3/8" at 1/32").
Use: 7X
Size: 26 mm