Learn More
Electron Microscopy Sciences Measuring Scale #6 for 7X Measuring Magnifier, 26 mm
SDP
Catalog No. 5027078
Encompass_Preferred
These scales are for use with the Measuring Magnifiers. Measuring scale for 7x magnifiers. Degrees: 0-90° (one 10° area marked in 1° intervals). Scales: ± 0 to 10mm (0.1mm divisions); 0 to 3/8" (1/64" divisions); 0 to .3" (.005" divisions). Radius: 0 to 180° (.5mm divisions).
Use: 7X
Size: 26 mm
Catalog No. 50-270-78 Supplier Electron Microscopy Sciences Supplier No. 68171
These scales are for use with the Measuring Magnifiers. Measuring scale for 7x magnifiers. Degrees: 0-90° (one 10° area marked in 1° intervals). Scales: ± 0 to 10mm (0.1mm divisions); 0 to 3/8" (1/64" divisions); 0 to .3" (.005" divisions). Radius: 0 to 180° (.5mm divisions).
Use: 7X
Size: 26 mm