Learn More
Electron Microscopy Sciences Measuring Scale #7 for 10X Measuring Magnifier, 35 mm
SDP
Supplier:  Electron Microscopy Sciences 68174
Encompass_Preferred
These scales are for use with the Measuring Magnifiers. Measuring scale for 10x magnifiers. Scale 7: Squares: 10 x 10mm (0.5mm divisions with 0.1mm markings). Circles: 0.1 to 2.5mm diameter.
Use: 10X
Size: 35 mm
Catalog No. 50-270-80