Learn More
Electron Microscopy Sciences Measuring Scale #7 for 7X Measuring Magnifier, 26 mm
SDP
Catalog No. 5027074
Encompass_Preferred
These scales are for use with the Measuring Magnifiers. Measuring scale for 7x magnifiers. Scale 7: Squares: 10 x 10mm (0.5mm divisions with 0.1mm markings). Circles: 0.1 to 2.5mm diameter.
Use: 7X
Size: 26 mm
Catalog No. 50-270-74 Supplier Electron Microscopy Sciences Supplier No. 68173
These scales are for use with the Measuring Magnifiers. Measuring scale for 7x magnifiers. Scale 7: Squares: 10 x 10mm (0.5mm divisions with 0.1mm markings). Circles: 0.1 to 2.5mm diameter.
Use: 7X
Size: 26 mm