Learn More
Electron Microscopy Sciences Measuring Scale #8 for 10X Measuring Magnifier, 35 mm
SDP
Catalog No. 5027087
Encompass_Preferred
These scales are for use with the Measuring Magnifiers. Measuring scale for 10x magnifiers. Scale: 0 to 20mm (0.1mm divisions).
Use: 10X
Size: 35 mm
Catalog No. 50-270-87 Supplier Electron Microscopy Sciences Supplier No. 68176
These scales are for use with the Measuring Magnifiers. Measuring scale for 10x magnifiers. Scale: 0 to 20mm (0.1mm divisions).
Use: 10X
Size: 35 mm