Learn More
Electron Microscopy Sciences Measuring Scale #8 for 7X Measuring Magnifier, 26 mm
SDP
Catalog No. 5027072
Encompass_Preferred
These scales are for use with the Measuring Magnifiers. Measuring scale for 7x magnifiers. Scale: 0 to 20mm (0.1mm divisions).
Use: 7X
Size: 26 mm
Catalog No. 50-270-72 Supplier Electron Microscopy Sciences Supplier No. 68175
These scales are for use with the Measuring Magnifiers. Measuring scale for 7x magnifiers. Scale: 0 to 20mm (0.1mm divisions).
Use: 7X
Size: 26 mm