Learn More
Electron Microscopy Sciences Measuring Scale #9 for 7X Measuring Magnifier, 26 mm
SDP
Catalog No. 5027081
Encompass_Preferred
These scales are for use with the Measuring Magnifiers. Measuring scale for 7x magnifiers. Degrees: 0-180° (1° intervals). Radius: 0 to 180°, 0 to 10mm (0.5mm divisions).
Use: 7X
Size: 26 mm
Catalog No. 50-270-81 Supplier Electron Microscopy Sciences Supplier No. 68177
These scales are for use with the Measuring Magnifiers. Measuring scale for 7x magnifiers. Degrees: 0-180° (1° intervals). Radius: 0 to 180°, 0 to 10mm (0.5mm divisions).
Use: 7X
Size: 26 mm