Learn More
BURKLE INC Measuring spoons 0.2 oz. (0.5 ml), PS, blue
Supplier: BURKLE INC 53784044
Blue tools support HACCP compliance by helping detect and prevent foreign object contamination—crucial in food processing, animal feed, and pharmaceutical industries. These volumetric and dosing spoons measure and dispense precise amounts of powders, granules, or liquids. Excess material can be leveled at the top edge, with volume engraved on the handle for quick reference. The flat front edge allows scooping from container bottoms and corners for complete emptying, while the flat base enables spill-free placement. Designed for single use, these disposable spoons are ideal for sampling small quantities, eliminating the need for cleaning and preventing cross-contamination or impurities. Made from high-quality plastic, they provide accuracy, safety, and convenience in laboratory, production, and quality control settings.