Learn More
Anton Paar Measuring Syste including cup and bob
Catalog No. 5000580506
Encompass_Preferred
Concentric cylinder systems when used with the corresponding cup are so called "absolute" measuring systems. This means that they have a defined shear gap and they measure the viscosity according to ISO 3219 and DIN 53019 (part 1). These standards describe the measurement of viscosity with rotational rheometers and define how to calculate the CSS and CSR factors of the "absolute" measuring systems
Catalog No. 50-005-80506 Supplier Anton Paar Supplier No. 20646
Concentric cylinder systems when used with the corresponding cup are so called "absolute" measuring systems. This means that they have a defined shear gap and they measure the viscosity according to ISO 3219 and DIN 53019 (part 1). These standards describe the measurement of viscosity with rotational rheometers and define how to calculate the CSS and CSR factors of the "absolute" measuring systems