Learn More
Anton Paar Measuring system DG26/SS - Stainless steel 26mm cup and bob double gap measuring system for ViscoQC 100 & ViscoQC 300
Catalog No. NC1818621
Encompass_Preferred
Measuring system consisting of bob B-DG26 and cup C-DG26/SS for measurement according to DIN 54453. The stainless steel hollow measuring bob is equipped with Toolmaster and magnetic coupling. A stainless steel hook connects the spindle body of the bob to the magnetic coupling. The cup consists of a stainless steel wall and bottom plug plus inner cylinder and an EPDM O-ring which seals the gap between wall and bottom.Requires holder for cup, either DIN Adapter item NC1818345 or PTD 80 item NC1818588Suitable for use with Anton Paar rotational viscometers ViscoQC 100 – L item NC1818559, ViscoQC 100 – R item NC1818567, ViscoQC 100 – H item NC1818569, ViscoQC 300 – L item NC1818572, ViscoQC 300 – R item NC1818576, ViscoQC 300 – H item NC1818578.
Catalog No. NC1818621 Supplier Anton Paar Supplier No. 223289
Measuring system consisting of bob B-DG26 and cup C-DG26/SS for measurement according to DIN 54453. The stainless steel hollow measuring bob is equipped with Toolmaster and magnetic coupling. A stainless steel hook connects the spindle body of the bob to the magnetic coupling. The cup consists of a stainless steel wall and bottom plug plus inner cylinder and an EPDM O-ring which seals the gap between wall and bottom.Requires holder for cup, either DIN Adapter item NC1818345 or PTD 80 item NC1818588Suitable for use with Anton Paar rotational viscometers ViscoQC 100 – L item NC1818559, ViscoQC 100 – R item NC1818567, ViscoQC 100 – H item NC1818569, ViscoQC 300 – L item NC1818572, ViscoQC 300 – R item NC1818576, ViscoQC 300 – H item NC1818578.