Learn More
Anton Paar Measuring system SC4-18 - Number 18 small sample cup and bob measuring system for ViscoQC 100 & ViscoQC 300
Catalog No. NC1818611
Encompass_Preferred
Measuring system consisting of bob B-SC4-18 and cup C-CC-18/SS for shear rate control and data analysis using mathematical models of software package V-Curve item NC1818583. The stainless steel measuring bob is equipped with Toolmaster and magnetic coupling. A stainless steel hook connects the spindle body of the bob to the magnetic coupling. The cup consists of a stainless steel wall, a PEEK bottom plug and an FKM O-ring which seals the gap between wall and bottom.Requires holder for cup, either DIN Adapter item NC1818345 or PTD 80 item NC1818588Suitable for use with Anton Paar rotational viscometers ViscoQC 100 – L item NC1818559, ViscoQC 100 – R item NC1818567, ViscoQC 100 – H item NC1818569, ViscoQC 300 – L item NC1818572, ViscoQC 300 – R item NC1818576, ViscoQC 300 – H item NC1818578.
Catalog No. NC1818611 Supplier Anton Paar Supplier No. 223516
Measuring system consisting of bob B-SC4-18 and cup C-CC-18/SS for shear rate control and data analysis using mathematical models of software package V-Curve item NC1818583. The stainless steel measuring bob is equipped with Toolmaster and magnetic coupling. A stainless steel hook connects the spindle body of the bob to the magnetic coupling. The cup consists of a stainless steel wall, a PEEK bottom plug and an FKM O-ring which seals the gap between wall and bottom.Requires holder for cup, either DIN Adapter item NC1818345 or PTD 80 item NC1818588Suitable for use with Anton Paar rotational viscometers ViscoQC 100 – L item NC1818559, ViscoQC 100 – R item NC1818567, ViscoQC 100 – H item NC1818569, ViscoQC 300 – L item NC1818572, ViscoQC 300 – R item NC1818576, ViscoQC 300 – H item NC1818578.