Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Measuring Worms™
SCI_ED

Catalog No. S78994
Change view
Click to view available options
Grade:
Pre-K to 12
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S78994 Pre-K to 12
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S78994 Supplier Learning Resources, Inc Supplier No. LER 0176
Add to Cart
Add to Cart

Includes

  • Worms in six colors and four sizes (1, 2, 3, and 4 in.)
  • Activity guide
  • Storage bucket

Worms anyone can love.

Students learn early math skills with these squishy worms that make counting, sorting and measuring fun.

Specifications

Grade Level Early Childhood, Elementary, Intermediate - Middle School, High School
Grade Pre-K to 12
Format Hands-on Learning
Product Type Measuring Worms™ Set
Includes Worms in six colors and four sizes 1, 2, 3 and 4 in., activity guide, storage bucket
For Use With (Application) Students learn early math skills with these squishy worms that make counting, sorting and measuring fun
Subject Mathematics
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.