Learn More
Med Vet International Dynarex Dynarule Bullseye Measuring Guide, 1250/Case
Catalog No. 502831653
Encompass_Preferred
Disposable, transparent wound measuring guide. Disposable, transparent wound measuring guide Straight rules are calibrated in inches and centimeters. Can measure up to 10cm. Flexible plastic sheets with bullseye grid and ruler. Permits easy length, width and diameter measurement of any wound.
Catalog No. 50-283-1653 Supplier Med Vet International Supplier No. 3007
Disposable, transparent wound measuring guide. Disposable, transparent wound measuring guide Straight rules are calibrated in inches and centimeters. Can measure up to 10cm. Flexible plastic sheets with bullseye grid and ruler. Permits easy length, width and diameter measurement of any wound.