Learn More
NEOGEN Meat Peptone No. 1, 500 g
SDP
Catalog No. 502015750
Encompass_Preferred
Meat Peptone No.1 is an enzymatic digest of animal tissue used in preparing microbiological culture media in a laboratory setting. Meat Peptone No.1 is not intended for use in the diagnosis of disease or other conditions in humans.Meat Peptone No. 1 was developed as an alternate nitrogen and vitamin source in dehydrated culture media to accommodate varying nutritional requirements.For more information visit https://www.neogen.com/solutions/Microbiology/meat-peptone-number-1
Catalog No. 50-201-5750 Supplier NEOGEN Supplier No. 700003526
Meat Peptone No.1 is an enzymatic digest of animal tissue used in preparing microbiological culture media in a laboratory setting. Meat Peptone No.1 is not intended for use in the diagnosis of disease or other conditions in humans.Meat Peptone No. 1 was developed as an alternate nitrogen and vitamin source in dehydrated culture media to accommodate varying nutritional requirements.For more information visit https://www.neogen.com/solutions/Microbiology/meat-peptone-number-1